Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H1.3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309327100UL
Description
Histone H1.3 Polyclonal specifically detects Histone H1.3 in Human samples. It is validated for Western Blot.Specifications
Histone H1.3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
H1 histone family, member 3, H1.3, H1F3H1D, histone 1, H1d, histone cluster 1, H1d, histone H1.3, Histone H1c, MGC138176 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Histone H1.3. Peptide sequence ETAPLAPTIPAPAEKTPVKKKAKKAGATAGKRKASGPPVSELITKAVAAS | |
100 μg | |
Epigenetics | |
3007 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction