Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1 Monoclonal Antibody (1415-1), Invitrogen™

Catalog No. PIMA513750
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513750 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513750 Supplier Thermo Scientific Supplier No. MA513750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MA5-13750 targets Histone H1 in IF, WB and IHC (P) applications and shows reactivity with human and mouse samples. The MA5-13750 immunogen is nuclei of myeloid leukemia biopsy cells.

This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1415-1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene H1-2
Gene Accession No. P07305, P10922, P15864, P16401, P16403, P43275, P43276, Q02539
Gene Alias 0610008C09Rik; CTL-1; H1; H1 antibody; H1 histone family member 0; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.0, H1(0), H1-0; H1.1; H1.12 linker histone, cluster member; H1.2; H1.2 linker histone, cluster member; H1.5; H1.9; H10; H1-0; H1-1; H1-12; H1-2; H1A; H1B; H1c; H1F0; H1f1; H1F2; H1F3; H1F5; H1FV; H1s-1; H1s-3; H1var1; H1var3; Hils1; His1a; HIST1; HIST1H1A; HIST1H1B; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1.1; histone H1.2; Histone H1.3; Histone H1.5; histone H1a; Histone H1b; histone H1c; Histone H1d; histone H1-like protein in spermatids 1; histone H1s-1; Histone H1s-2; Histone H1s-3; histone linker H1 domain, spermatid-specific 1 (pseudogene); spermatid-specific linker histone H1-like protein; TISP64
Gene Symbols H1-0, H1-1, H1-2, H1-5
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14958, 3005, 3006, 3009, 3024, 50708, 56702, 80838
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.