Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Histone H1T Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566793 Supplier Invitrogen™ Supplier No. PA566793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RATTPKTVRSG RKAKGAKGKQ QQKSPVKARA SKSKLTQHHE VNVRKATSKK Highest antigen sequence identity to the following orthologs - mouse 48%, rat 48%.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1T
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene H1-6
Gene Accession No. P22492
Gene Alias dJ221C16.2; H1 histone family, member T (testis-specific); H1.6; H1ft; H1T; HIST1H1T; histone 1, H1t; histone cluster 1, H1t; histone H1t; Testicular H1 histone
Gene Symbols H1-6
Host Species Rabbit
Immunogen Recombinant Human Histone H1T. Recombinant protein control fragment (Product #RP-107450).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.