Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1t Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502279386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9386 Supplier Bioss Supplier No. BS1413RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H1t Polyclonal antibody specifically detects Histone H1t in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Histone H1t
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene H1-6
Gene Accession No. P22492, Q07133
Gene Alias dJ221C16.2; H1 histone family, member T (testis-specific); H1.6; H1ft; H1T; HIST1H1T; histone 1, H1t; histone cluster 1, H1t; histone H1t; Testicular H1 histone
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Testicular H1 histone (51-207).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107970, 3010
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.