Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H1T Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $389.00
Specifications
Antigen | Histone H1T |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Histone H1T Polyclonal antibody specifically detects Histone H1T in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Histone H1T | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
dJ221C16.2, H1 histone family, member T (testis-specific), H1FTMGC163222, H1t, histone 1, H1t, histone cluster 1, H1t, histone H1t, Testicular H1 histone | |
A synthetic peptide corresponding to a sequence within amino acids 150-209 of mouse Histone H1T (NP_034507.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Epigenetics | |
PBS, 50% glycerol, pH7.3 | |
3010 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title