Learn More
Histone H1x Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304604AT
 
            Description
The recommended shelf life for this product is 1 year from date of receipt.
Histones H1 are necessary for the condensation of nucleosome chains into higher-order structures.Specifications
| Histone H1x | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q92522 | |
| H1FX | |
| Between 163 and 213 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
| Western Blot | |
| Unconjugated | |
| H1FX | |
| Histone H1x, H1FX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8971 | |
| 4° C | |
| Liquid | 
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
