Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A-Bbd type 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $501.50

Specifications

Antigen Histone H2A-Bbd type 1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124124
SDP
View Documents
Novus Biologicals
NBP30961125UL
25 μg
Each for $206.00
Only null left
Add to Cart
 
NB124123
SDP
View Documents
Novus Biologicals
NBP309611100UL
100 μg
Each for $501.50
Only null left
Add to Cart
 
Description

Description

Histone H2A-Bbd type 1 Polyclonal specifically detects Histone H2A-Bbd type 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Histone H2A-Bbd type 1
Western Blot
Unconjugated
Rabbit
Human
H2A.Bbd
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Histone H2A-Bbd type 1 (NP_001017990). Peptide sequence SSGAGGRGRTCSRTVRAELSFSVSQVERSLREGHYAQRLSRTAPVYLAAV
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
474382
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.