Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2A (hydroxyl Y39) Rabbit anti-Human, Unconjugated, Clone: DED-8, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20142
Description
Histone H2A (hydroxyl Y39) Monoclonal antibody specifically detects Histone H2A (hydroxyl Y39) in Human samples. It is validated for WB, IHC-P.Specifications
Histone H2A (hydroxyl Y39) | |
Monoclonal | |
Unconjugated | |
A synthetic hydroxyl peptide corresponding to residues surrounding Y39 of Histone H2A | |
Primary | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
DED-8 | |
Rabbit | |
100 μg | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction