Learn More
Histone H2A.J Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H2A.J |
| Applications | Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | H2AFJ |
| Gene Accession No. | Q8R1M2, Q9BTM1 |
| Gene Alias | buforin I; H2A histone family member J; H2A histone family, member J; H2a/j; H2AFJ; H2AJ; Histone H2A.J |
| Gene Symbols | H2AFJ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.