Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2a, p Ser129 Antibody (SR1183), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32152650UL
Description
Histone H2a Monoclonal antibody specifically detects Histone H2a in Yeast samples. It is validated for Western BlotSpecifications
Histone H2a | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Yeast | |
Purified |
Western Blot | |
SR1183 | |
Western Blot 1:1000-1:2000 | |
H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460 | |
A synthesized peptide derived from yeast phospho-Histone H2a [Ser129] (Uniprot #: P04912) | |
50 μg | |
Chromatin Research, Epigenetics | |
8338 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction