Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992987
Encompass
Change view
Click to view available options
Quantity:
10 μL
10 μg
100 μL
100 μg
50 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2987 100 μg
50-199-2988 10 μg
50-199-3955 10 μL
50-227-7242 50 μL
50-199-3954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-199-2987 Supplier Active Motif Supplier No. 39945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

Specifications

Antigen Histone H2A
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Alias 1190022L06; Acetyl-Histone H2; AW228881; BOS_14476; BOS_21793; BOS_6451; dJ160A22.4; dJ86C11.1; gammaH2ax; h2a; H2A 615 antibody; H2A GL101 antibody; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1 antibody; H2A.1b; H2A.2; H2A.3; H2a.x; H2A.z; H2a.z-1; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/S; H2a/x; H2A/z; H2a-291A; H2a-53; h2a-A; H2ac11; H2ac13; H2AC15; H2AC16; H2AC17; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2afx; H2AFZ; H2AG; H2AH; H2ak; H2AK5ac; H2ax; H2AX histone; H2az; H2AZ histone; HIS; Hist1h2aa; Hist1h2ab; Hist1h2ac; HIST1H2AD; HIST1H2AE; HIST1H2AE antibody; HIST1H2AG; Hist1h2ah; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1
Host Species Rabbit
Immunogen This Histone H2A antibody was raised against recombinant yeast histone H2A.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.