Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A Rabbit anti-Human, Clone: RM225, Abnova™

Catalog No. 89035326 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-326 Supplier Abnova Corporation Supplier No. MAB15118
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic peptide of human histone H2A.

  • High specificity
  • High reactivity

Specifications

Antigen Histone H2A
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RM225
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human histone H2A.
Dilution ELISA (0.2-1 ug/mL) Immunocytochemistry (1-2 ug/mL) Multiplex (0.2-1 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P04908; Q93077; Q7L7L0; Q6FI13; P20671; P0C0S8
Gene Alias H2A.1/H2A.2/H2A/a/H2AFA
Gene Symbols HIST1H2AE
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human histone H2A.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3012/8334/95815/723790/3013/8329
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.