Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2A.Z Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298970AF350
Description
Histone H2A.Z Polyclonal specifically detects Histone H2A.Z in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Histone H2A.Z | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 350 | |
H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Histone H2A.Z. | |
0.1 mL | |
Epigenetics | |
3015 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction