Learn More
Invitrogen™ Histone H2A.Z Polyclonal Antibody

Description
Recommended positive controls: Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Dog (100%), Pig (100%), Chicken (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Histone H2A.Z |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | H2AZ1 |
| Gene Accession No. | P0C0S5, P0C0S6, Q3THW5, Q71UI9 |
| Gene Alias | 11ac; C530002L11Rik; H2A histone family member V; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Va; H2A histone family, member Z; H2A.F/Z; H2A.z; H2A.Z variant histone 1; H2A.Z variant histone 2; H2A.Z variant histone 2a; H2A.Z1; H2a.z-1; H2A.Z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2AF; H2AFV; h2afva; h2afz; h2afz.L; H2AFZac; H2AV; H2az; H2AZ histone; H2AZ1; H2AZ2; h2az2a; h2aza; H2AZK11ac; H2AZK7ac; histone (H2A.Z); histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; histone H2A.Z-like; histone variant H2A.F/Z; hypothetical protein; purine-rich binding element protein B; RCJMB04_2h11; Unknown (protein for MGC:133654); unnamed protein product; XELAEV_18005732mg; XH2AZ |
| Gene Symbols | H2AFZ, H2AZ1, H2az2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.