Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.Z Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993886
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3886 10 μL
50-227-7220 50 μL
50-199-3885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-3886 Supplier Active Motif Supplier No. 39114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.

Specifications

Antigen Histone H2A.Z
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene H2AZ1
Gene Accession No. P0C0S5
Gene Alias 11ac; BOS_6451; C530002L11Rik; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Z; H2A.F/Z; H2A.Z; H2A.Z1; H2A.Z-1; H2a.z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2afv; H2afz; h2afz.L; H2AFZac; H2av; H2AZ; H2AZ histone; H2AZK11ac; H2AZK7ac; histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; Histone H2A.Z-like; XELAEV_18005732mg; XH2AZ
Gene Symbols H2AZ1
Host Species Rabbit
Immunogen This Histone H2A.Z antibody was raised against a peptide derived from the C-terminus of human histone H2A.Z.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3015
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.