Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Histone H2A.Z Recombinant Rabbit Monoclonal Antibody (ST46-09)
GREENER_CHOICE

Catalog No. PIMA541132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541132 Supplier Invitrogen™ Supplier No. MA541132
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is predicted to work in Western Blot at a dilution of 1:500-1:1,000.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.Z
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone ST46-09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene H2AZ1
Gene Accession No. P0C0S5, P0C0S6, P0C0S7, Q3THW5, Q71UI9
Gene Alias 11ac; C530002L11Rik; H2A histone family member V; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Va; H2A histone family, member Z; H2A.F/Z; H2A.z; H2A.Z variant histone 1; H2A.Z variant histone 2; H2A.Z variant histone 2a; H2A.Z1; H2a.z-1; H2A.Z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2AF; H2AFV; h2afva; h2afz; h2afz.L; H2AFZac; H2AV; H2az; H2AZ histone; H2AZ1; H2AZ2; h2az2a; h2aza; H2AZK11ac; H2AZK7ac; histone (H2A.Z); histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; histone H2A.Z-like; histone variant H2A.F/Z; hypothetical protein; purine-rich binding element protein B; RCJMB04_2h11; Unknown (protein for MGC:133654); unnamed protein product; XELAEV_18005732mg; XH2AZ
Gene Symbols H2AFZ, H2AZ1, H2az2
Host Species Rabbit
Immunogen Synthetic peptide within human Histone H2AZ aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3015, 51788, 58940, 77605, 94239
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.