Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AE Antibody (RM225), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326042AF647
Description
Histone H2AE Monoclonal antibody specifically detects Histone H2AE in Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex ImmunoassaySpecifications
Histone H2AE | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Vertebrates | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Multiplex | |
RM225 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay | |
H2A histone family, member M, H2A/m, H2AFA, H2AFMHistone H2A/m, HIST1H2AE, histone 1, H2ab, histone cluster 1, H2ab, histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a | |
A peptide corresponding to the C-terminus of Human Histone H2AE | |
0.1 mL | |
Epigenetics | |
3012 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction