Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AK9ac Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993882
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3882 10 μL
50-199-3881 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3882 Supplier Active Motif Supplier No. 39110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

Specifications

Antigen Histone H2AK9ac
Applications Dot Blot, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene H2AC12
Gene Accession No. P04908, P0C0S5, P0C0S8, P16104, P20671, Q96KK5, Q99878
Gene Alias 1190022L06; Acetyl-Histone H2; Acetyl-Histone H2A K9; Acetyl-Histone H2A Lys9; AW228881; dJ160A22.4; dJ86C11.1; gammaH2ax; H2A 615; H2A GL101; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1b; H2A.2; H2A.3; H2a.x; H2A.Z; H2A.Z-1; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/S; H2a/x; H2A/z; H2a-291A; H2a-53; H2ac11; H2ac13; H2AC15; H2AC16; H2AC17; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2afx; H2AFZ; H2AG; H2AH; H2ak; H2AK5ac; H2ALys9ac; H2ax; H2AX histone; H2az; H2AZ histone; HIS; Hist1h2aa; Hist1h2ab; Hist1h2ac; HIST1H2AD; HIST1H2AE; HIST1H2AG; Hist1h2ah; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1h2an; Hist1h2ao; Hist1h2ap; Hist1h2ba; HIST3H2A; Hist5-2ax; histone 1, H2ab; histone 1, H2ad; histone 1, H2ae; histone 1, H2ag; histone 1, H2ah; histone 1, H2ai; histone 1, H2aj; histone 1, H2ba; histone 2, H2ai; histone 5 protein 2ax; histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2aj; histone cluster 1, H2ba; Histone H2A type 1; Histone H2A type 1-B; Histone H2A type 1-B/E; Histone H2A type 1-C; Histone H2A type 1-D; Histone H2A type 1-E; Histone H2A type 1-G; Histone H2A type 1-H; Histone H2A type 1-I; Histone H2A type 1-J; Histone H2A type 1-N; Histone H2A type 1-O; Histone H2A type 1-P; Histone H2A type 4; Histone H2A, testis; Histone H2A.2; Histone H2A.3; Histone H2A.X; Histone H2A.Z; Histone H2A/a; Histone H2A/e; Histone H2A/g; Histone H2A/m; histone H2A/p; Histone H2A/ptl; Histone H2A/s; histone H2AD; histone H2AE; Histone H2AX; Histone H2B type 1-A; Histone H2B, testis; MGC74460; pH2A/f; RGD1566119; testis-specific histone 2a; testis-specific histone 2b; Testis-specific histone H2B; TH2A; Th2b
Gene Symbols H2AC11, H2AC12, H2AC13, H2AC14, H2AC4, H2AC7, H2AC8, H2AX, H2AZ1
Host Species Rabbit
Immunogen This Histone H2A acetyl Lys9 antibody was raised against a peptide including acetyl-lysine 9 of human histone H2A.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3012, 3013, 3014, 3015, 8329, 8331, 8335, 85235, 8969
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.