Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 11 publications
Supplier: Novus Biologicals NB100383
Description
Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Histone H2AX | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
H2A.X, H2A/X, H2AFX | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for H2AX. It is not known if this also recognizes the phosphorylated form of H2AX. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Simple Western 10 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500-1:2000 | |
P16104 | |
H2AFX | |
The immunogen recognized by this antibody maps to the C-terminus of human histone H2AX using the numbering given for Swiss-Prot entry P16104 (GeneID 3014). | |
100 μL | |
DNA Repair, Mitotic Regulators | |
3014 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction