Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AX, p Ser139 Antibody (2207D), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2288R
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Histone H2AX | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
2207D | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
H2A.X, H2A/X, H2AFX | |
Phospho-peptide containing human Histone H2AX S139 site, Accession # P16104 | |
0.1 mL | |
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
3014 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction