Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AX, p Ser139 Antibody (RM224), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP318533
This item is not returnable.
View return policy
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human, Vertebrates samples. It is validated for Western Blot, ELISA, Immunofluorescence, Multiplex ImmunoassaySpecifications
| Histone H2AX | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 0.5 to 2 μg/mL, ELISA 0.2 to 1 μg/mL, Immunocytochemistry/Immunofluorescence 0.5 to 2 μg/mL, Multiplex Immunoassay 0.1 to 1 μg/mL | |
| H2A.X, H2A/X, H2AFX | |
| A phospho-peptide corresponding to Phospho-Histone H2AX (Ser139) | |
| 100 μg | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
| 3014 | |
| Store at -20°C. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Peptide Array | |
| RM224 | |
| Unconjugated | |
| 50% Glycerol, PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction