Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX (phospho S139) Rabbit anti-Human, Clone: RM224, Abnova™

Catalog No. 89035319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-319 Supplier Abnova Corporation Supplier No. MAB15111
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic phosphopeptide of human histone H2AX (S139).

  • High specificity
  • High reactivity

Specifications

Antigen Histone H2AX (phospho S139)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RM224
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic phosphopeptide of human histone H2AX (S139).
Dilution ELISA (0.2-1 ug/mL) Immunocytochemistry (0.5-2 ug/mL) Multiplex (0.1-1 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P16104
Gene Alias H2A.X/H2A/X/H2AX
Gene Symbols H2AFX
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S139 of human histone H2AX.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity This antibody reacts to histone H2AX only when phosphorylated at S139. No cross reactivity with other phosphorylated histones.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.