Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093218 20 μL
NB093217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093218 Supplier Novus Biologicals Supplier No. NBP30395020UL

Rabbit Polyclonal Antibody

Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Histone H2AX
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human Histone H2AX (NP_002096.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.