Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX Rabbit, Clone: RM214, Abnova™

Catalog No. 89934248 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-248 Supplier Abnova Corporation Supplier No. MAB12802
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of human Histone H2AX.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif. [provided by RefSeq]

Specifications

Antigen Histone H2AX
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RM214
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human Histone H2AX.
Dilution ELISA (0.2 ug/mL-1 ug/mL) Immunocytochemistry (1 ug/mL-2 ug/mL) Western Blot (0.5 ug/mL-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias H2A.X/H2A/X/H2AX
Gene Symbols H2AFX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of Histone H2AX.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity This antibody reacts to Histone H2AX protein, independent of post-translational modifications. No cross reactivity with other histone proteins. .
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.