Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AY/macroH2A.1 Antibody (CL5256), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Histone H2AY/macroH2A.1 |
---|---|
Clone | CL5256 |
Dilution | Immunohistochemistry 1:200 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Histone H2AY/macroH2A.1 Monoclonal specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Histone H2AY/macroH2A.1 | |
Immunohistochemistry 1:200 - 1:500 | |
Unconjugated | |
Mouse | |
core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 | |
H2AFY | |
IgG1 | |
Protein A purified |
CL5256 | |
Monoclonal | |
Purified | |
Human | |
9555 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GEVSKAASADSTTEGTPA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title