Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Histone H2B.2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548430
Description
Predicted to react with mouse based on sequence homology.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
Histone H2B.2 | |
Polyclonal | |
Unconjugated | |
H2BC17 | |
dJ193B12.2; H2B histone family, member N; H2B.2; H2B/n; H2BFH; H2BFN; HIST1H2BO; histone 1, H2bo; histone cluster 1, H2bo; histone H2B type 1-O; Histone H2B.2; histone H2B.n; Histone H2B2; Histone H2Bn | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P23527 | |
H2BC17 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HIST1H2BO/HIST1H2BH. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction