Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B, ac Lys11 Antibody (RM456), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326035AF700
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex ImmunoassaySpecifications
Histone H2B | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Vertebrates | |
Purified |
Western Blot, ELISA, Multiplex | |
RM456 | |
Western Blot, ELISA, Multiplex Immunoassay | |
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
An acetyl-peptide corresponding to Histone H2B [Lys11] (Uniprot # P62807) | |
0.1 mL | |
Chromatin Research, Epigenetics | |
440689 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction