Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B (acetyl K20) Rabbit anti-Human, Clone: RM235, Abnova™

Catalog No. 89035320 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-320 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-320 Supplier Abnova Corporation Supplier No. MAB15112
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic acetyl peptide of human histone H2B (acetyl K20).

  • High specificity
  • High reactivity

Specifications

Antigen Histone H2B (acetyl K20)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RM235
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic acetyl peptide of human histone H2B (acetyl K20).
Dilution ELISA (0.2-1 ug/mL) Immunocytochemistry (1-2 ug/mL) Multiplex (0.1-0.5 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P33778
Gene Alias H2B.1/H2B/f/H2BFF/MGC119804
Gene Symbols HIST1H2BB
Host Species Rabbit
Immunogen A synthetic acetyl peptide corresponding to residues surrounding K20 of human histone H2B.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3018
Test Specificity This antibody reacts to histone H2B acetylated at K20. No cross reactivity with non-modified K20 or other acetylated lysines in histone H2B.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.