Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Polyclonal Antibody, Active Motif™
SDP

Catalog No. 502277243
Encompass
Change view
Click to view available options
Quantity:
10 μL
10 μg
100 μL
100 μg
50 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7243 50 μL
50-199-2990 10 μg
50-199-2989 100 μg
50-199-3957 10 μL
50-199-3956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 50-227-7243 Supplier Active Motif Supplier No. 39437
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.

Specifications

Antigen Histone H2B
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene Alias 2610022J01Rik; Acetyl-Histone H2; AV127319; BOS_21303; BOS_21904; BOS_2944; BOS_2955; CG17949; CG17949 gene product from transcript CG17949-RA; CG17949-PA; CG33868; CG33870; CG33872; CG33874; CG33876; CG33878; CG33880; CG33882; CG33884; CG33886; CG33888; CG33890; CG33892; CG33894; CG33896; CG33898; CG33900; CG33902; CG33904; CG33906; CG33908; CG33910; dJ193B12.2; dJ221C16.3; dJ221C16.8; Dmel\CG17949; Dmel_CG17949; fj05g08; GL105; H2B; H2b 613; H2B GL105 antibody; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B I; H2B II; H2B III; H2B IV; H2B K; H2B VI; H2B VII; H2B.1; H2B.1 antibody; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/S; H2B/t; H2B1.1; H2b-613; H2BC10; H2BC4; H2bc6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; h2bfr; H2bfs; H2BFT; H2BGL105; H2B-I; H2B-II; H2B-III; H2B-IV; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BQ; H2BS14; H2BS14p; H2B-VI; H2B-VII; HIRA-interacting protein 1; HIRIP1; His2B; His2B:CG17949; His2B:CG17949-PA; His2B:CG33868; His2B:CG33870; His2B:CG33872; His2B:CG33874; His2B:CG33876; His2B:CG33878; His2B:CG33880; His2B:CG33882; His2B:CG33884; His2B:CG33886; His2B:CG33888; His2B:CG33890; His2B:CG33892; His2B:CG33894; His2B:CG33896; His2B:CG33898; His2B:CG33900; His2B:CG33902; His2B:CG33904; His2B:CG33906; His2B:CG33908; His2B:CG33910; HIST1H2B7; Hist1h2ba; HIST1H2BB; HIST1H2BB antibody; HIST1H2BC; HIST1H2BE; HIST1H2BF; Hist1h2bg; HIST1H2BI; hist1h2bj; hist1h2bj.L; hist1h2bj.S; Hist1h2bk; HIST1H2BM; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; Hist2h2be; HIST2H2BF; hist2h2l; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2, like; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bj L homeolog; histone cluster 1, H2bj S homeolog; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 1, H2B-VII (similar to human histone cluster 1, class H2B, member N); histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; Histone H2B; Histone H2B 1.1; Histone H2B 1/2/3/4/6; Histone H2B 3; histone H2B 7; Histone H2B type 1; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; histone H2B-like; LOC608682; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b; Unknown (protein for MGC:73093); wu:fj05g08; XELAEV_18000725mg; zgc:73093
Host Species Rabbit
Immunogen This yeast histone H2B antibody was raised against recombinant yeast histone H2B.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.