Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Rabbit anti-Human, Clone: 3A6, Bioss
SDP

Catalog No. 501989578
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9578 Supplier Bioss Supplier No. BSM52099R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.

Specifications

Antigen Histone H2B
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A6
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene H2BC12
Gene Accession No. O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q96A08, Q99877
Gene Alias 2610022J01Rik; Acetyl-Histone H2; AV127319; BOS_21303; BOS_21904; BOS_2944; BOS_2955; CG17949; CG17949 gene product from transcript CG17949-RA; CG17949-PA; CG33868; CG33870; CG33872; CG33874; CG33876; CG33878; CG33880; CG33882; CG33884; CG33886; CG33888; CG33890; CG33892; CG33894; CG33896; CG33898; CG33900; CG33902; CG33904; CG33906; CG33908; CG33910; dJ193B12.2; dJ221C16.3; dJ221C16.8; Dmel\CG17949; Dmel_CG17949; fj05g08; GL105; H2B; H2b 613; H2B GL105 antibody; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B I; H2B II; H2B III; H2B IV; H2B K; H2B VI; H2B VII; H2B.1; H2B.1 antibody; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/S; H2B/t; H2B1.1; H2b-613; H2BC10; H2BC4; H2bc6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; h2bfr; H2bfs; H2BFT; H2BGL105; H2B-I; H2B-II; H2B-III; H2B-IV; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BQ; H2BS14; H2BS14p; H2B-VI; H2B-VII; HIRA-interacting protein 1; HIRIP1; His2B; His2B:CG17949; His2B:CG17949-PA; His2B:CG33868; His2B:CG33870; His2B:CG33872; His2B:CG33874; His2B:CG33876; His2B:CG33878; His2B:CG33880; His2B:CG33882; His2B:CG33884; His2B:CG33886; His2B:CG33888; His2B:CG33890; His2B:CG33892; His2B:CG33894; His2B:CG33896; His2B:CG33898; His2B:CG33900; His2B:CG33902; His2B:CG33904; His2B:CG33906; His2B:CG33908; His2B:CG33910; HIST1H2B7; Hist1h2ba; HIST1H2BB; HIST1H2BB antibody; HIST1H2BC; HIST1H2BE; HIST1H2BF; Hist1h2bg; HIST1H2BI; hist1h2bj; hist1h2bj.L; hist1h2bj.S; Hist1h2bk; HIST1H2BM; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; Hist2h2be; HIST2H2BF; hist2h2l; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2, like; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bj L homeolog; histone cluster 1, H2bj S homeolog; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 1, H2B-VII (similar to human histone cluster 1, class H2B, member N); histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; Histone H2B; Histone H2B 1.1; Histone H2B 1/2/3/4/6; Histone H2B 3; histone H2B 7; Histone H2B type 1; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; histone H2B-like; LOC608682; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b; Unknown (protein for MGC:73093); wu:fj05g08; XELAEV_18000725mg; zgc:73093
Gene Symbols H2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8, HIST1H2BA
Host Species Rabbit
Immunogen Recombinant human Histone H2B protein, full length.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255626, 337873, 337874, 440689, 8339, 8341, 8346, 8347, 8348, 8349, 85236
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.