Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259270
Description
Histone H2B Polyclonal specifically detects Histone H2B in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
| Histone H2B | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000, Dot Blot, Chromatin Immunoprecipitation (ChIP) 0.5-1 ug/IP | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
| HIST2H2BF | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| 440689 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction