Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B, ubiquitylated Lys123 Antibody (1B3F12/A9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254770AF532
Description
Histone H2B Monoclonal specifically detects Histone H2B in Human, S. pombe, Yeast samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Histone H2B | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
HIST2H2BF | |
Ub-polypeptide antigen based on amino acids 115 - 130 of yeast Histone H2B | |
0.1 mL | |
Primary | |
Human, Yeast | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1B3F12/A9 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
Mouse | |
Protein A purified | |
Chromatin Research, Epigenetics, Histones and Modified Histones | |
440689 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction