Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2BK15ac Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993328
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3328 100 μL
50-199-3329 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3328 Supplier Active Motif Supplier No. 61321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.

Specifications

Antigen Histone H2BK15ac
Applications Dot Blot, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene H2BC12
Gene Accession No. O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q99877
Gene Alias 2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K15; Acetyl-Histone H2B Lys15; AV127319; dJ193B12.2; dJ221C16.3; dJ221C16.8; GL105; H2B; H2b 613; H2B GL105; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B K; H2B.1; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/s; H2B/t; H2b-613; H2BC10; H2BC4; H2BC6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; H2bfs; H2BFT; H2BGL105; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BLys15ac; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRIP1; Hist1h2ba; Hist1h2bc; Hist1h2be; HIST1H2BF; HIST1H2BG; HIST1H2BI; Hist1h2bk; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; HIST2H2BE; HIST2H2BF; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; histone H2B; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b
Gene Symbols H2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8
Host Species Rabbit
Immunogen This antibody was raised against a peptide including acetyl-lysine 15 of human histone H2B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337873, 337874, 440689, 8339, 8341, 8346, 8347, 8348, 8349, 85236
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.