Learn More
Histone H2BK46ac Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Histone H2BK46ac |
| Applications | ChIP Assay, Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Gene | HIST1H2BA |
| Gene Accession No. | Q96A08, Q96QV6 |
| Gene Symbols | HIST1H2AA, HIST1H2BA |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.