Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2BK5me1 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
Antigen | Histone H2BK5me1 |
---|---|
Concentration | Lot-Specific |
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3399
![]() ![]() |
Active Motif
61440 |
10 μL |
Each for $159.90
|
|
|||||
50-199-3398
![]() ![]() |
Active Motif
61439 |
100 μL |
Each for $633.45
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.Specifications
Histone H2BK5me1 | |
Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96A08, Q96QV6 | |
HIST1H2AA, HIST1H2BA | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
221613, 255626 | |
This antibody was raised against a synthetic peptide containing monomethyl Lysine 5 of human histone H2B. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
HIST1H2BA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title