Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3.1, ac Lys14 Antibody (3H3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Histone H3.1 |
---|---|
Clone | 3H3 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:500 |
Applications | Western Blot, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
Histone H3.1 | |
Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
Synthesized peptide [UniProt P68431] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3H3 | |
Western Blot, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Epigenetics, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
8353 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title