Learn More
Description
Specifications
Specifications
| Antigen | Histone H3.1 |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 |
| Formulation | phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide |
| Gene Accession No. | P68431 |
| Gene Alias | H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l |
| Gene Symbols | HIST1H3E |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
