Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3.1, Methyl Lys79 Antibody (2G10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32613650UL
Description
Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human, Rat samples. It is validated for ELISASpecifications
Histone H3.1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
ELISA | |
2G10 | |
ELISA | |
H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
Synthesized peptide. The Histone H3.1 [Methyl Lys79] Recombinant Monoclonal Antibody was raised against a mixture of peptides containing 1, 2 or 3 methyl groups, such as DF-(Me1)K-TD and DF-(Me2)K-TD and DF-(Me3)K-TD. [UniProt P68431] | |
50 μL | |
Epigenetics, Phospho Specific | |
8353 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction