Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3.3 Antibody (4H2), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326227100UL
Description
Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Histone H3.3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry | |
4H2 | |
ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 | |
H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783 | |
Synthesized peptide [UniProt P84243] | |
100 μL | |
Chromatin Research, Epigenetics | |
3020 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction