Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H3F3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560027 Supplier Invitrogen™ Supplier No. PA560027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEAYLVGLFE DTNLCAIHAK RVTIM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H3F3A
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene H3-3A
Gene Accession No. P84243
Gene Alias H3 histone, family 3A; H3.3A; H3.3B; H3/a; H3-3A; H3-3B; H3F3; H3F3A; H3F3B; Histone 3; Histone H3.3; PP781
Gene Symbols H3-3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Histone H3.3. Recombinant protein control fragment (Product #RP-104343).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.