Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Histone H3.3 Recombinant Rabbit Monoclonal Antibody (RM190)
GREENER_CHOICE

Catalog No. PIMA524667
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524667 Supplier Invitrogen™ Supplier No. MA524667
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody reacts to Histone H3.3, independent of post-translational modifications. No cross reactivity with Histone H3.1 or other histone proteins. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.3
Applications ChIP Assay, ELISA, Western Blot, Luminex
Classification Recombinant Monoclonal
Clone RM190
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.09% sodium azide; pH 7.2-7.4
Gene H3-3A
Gene Accession No. P84243
Gene Alias 9430068D06Rik; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; family 3B (H3.3B); H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 histone A; H3.3 histone B; H3.3A; H3.3A/B; H3.3B; H3.4; H3.A; H3.B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3A protein; H3F3B; H3FT; H3-IX; H3L-like histone; H3t; H3-X; His3.3; His3.3A; His-3.3A; His3.3A-PA; His3.3A-PC; His3.3A-PD; His3.3B; histone; Histone cluster 3; Histone H3; histone H3 class II; Histone H3.3; histone H3.3A; Histone H3.3B; histone H3.3-like protein; MGC126886; MGC126888; PH3; PP781; similar to H3 histone, family 3A; unnamed protein product
Gene Symbols H3-3A, H3-3B
Host Species Rabbit
Immunogen Peptide corresponding to human Histone H3.3.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3020, 3021
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.