Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, ac Lys14 Antibody (RM130) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325982
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), Multiplex ImmunoassaySpecifications
Histone H3 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Vertebrates | |
Purified |
Western Blot, ELISA, Immunocytochemistry, ChIP Assay, Multiplex | |
RM130 | |
Western Blot : 0.5 ug/ml-2 ug/ml, ELISA : 0.2 ug/mL- 1 ug/mL, Immunocytochemistry/ Immunofluorescence : 0.5 ug/ml - 2 ug/ml, Chromatin Immunoprecipitation (ChIP) : 2 ug/mL- 10 ug/mL, Multiplex Immunoassay : 0.1 ug/ml - 0.5 ug/ml | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
This Histone H3 [ac Lys14] antibody (RM130) was raised against an acetyl-peptide corresponding to the Histone H3 [ac Lys14] | |
100 μg | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction