Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3, ac Lys36 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB211254

 View more versions of this product

Catalog No. NB211254


Only null left
Add to Cart

Description

Description

Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Dot Blot.
Specifications

Specifications

Histone H3 (ac Lys36)
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Rabbit
15 kDa
0.05 mg
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Human, Mouse, C. elegans
IgG
Western Blot, Dot Blot
0.47 mg/mL
Western Blot 1:1000, Dot Blot 1:1000
Q71DI3, P84228
H3C14
This Histone H3 [ac Lys36] antibody was raised against synthetic acetylated peptide surrounding Lysine 36 of human Histone H3.2 [Swiss Prot Q71DI3].
Affinity Purified
RUO
Primary
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only