Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, ac Lys56 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259214
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot, Chromatin Immunoprecipitation Sequencing.Specifications
Histone H3 (ac Lys56) | |
Polyclonal | |
Western Blot, ELISA 1:500, Immunocytochemistry/Immunofluorescence, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 5 ug/IP, Chromatin Immunoprecipitation Sequencing | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K56ac, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
50 μg | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot | |
Unconjugated | |
PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
H3C14 | |
The exact sequence of the immunogen to this Histone H3 [ac Lys56] antibody is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction