Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, ac Lys9, ac Lys14 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$243.50 - $402.00
Specifications
Antigen | Histone H3 (ac Lys9, ac Lys14) |
---|---|
Concentration | 0.63 mg/mL |
Dilution | Western Blot 1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 0.1 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Dot Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB211081SS
![]() |
Novus Biologicals
NB211081SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB211081
![]() |
Novus Biologicals
NB211081 |
0.05 mg |
Each for $402.00
|
|
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
Histone H3 (ac Lys9, ac Lys14) | |
Western Blot 1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:200, Dot Blot 0.1 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
Polyclonal | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9ac, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
H3C14 | |
IgG | |
Affinity Purified | |
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. |
0.63 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Dot Blot | |
Unconjugated | |
RUO | |
Human, Mouse, C. elegans | |
Q71DI3, P84228 | |
126961 | |
This Histone H3 [ac Lys9, ac Lys14] antibody was raised against synthetic acetylated peptide surrounding Lysines 9 and 14 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title