Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Histone H3 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Histone H3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
H3C14 | |
IgG | |
Affinity Purified | |
15 kDa |
Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
Polyclonal | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
126961 | |
This Histone H3 antibody was developed against a Recombinant Protein corresponding to amino acids: SEAYLVGLFEDTNLCAIHAKRVTIM | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title