Learn More
Histone H3, Asym-dimethyl Arg9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244066
Description
Specifically detects Histone H3 (Asym-dimethyl Arg9) in C. elegans, Chicken, Drosophila, Human, Mouse, Plant, Rat, Xenopus samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Histone H3 (Asym-dimethyl Arg9) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with 0.01% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3R9Me2a, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
0.05 mg | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
0.84 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Chromatin Immunoprecipitation (ChIP) 2-5ug/million cells | |
Q71DI3 | |
H3C14 | |
Histone H3 [Asym-dimethyl Arg9] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic dimethylated peptide surrounding Arginine 8 of human Histone H3.2. (Uniprot: Q71DI3) | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, C. elegans, Chicken, Drosophila, Drosophila, Plant, Xenopus species | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only