Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Citrulline Arg17, Citrulline Arg2, Citrulline Arg8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10057135
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Histone H3 (Citrulline Arg17, Citrulline Arg2, Citrulline Arg8) | |
Polyclonal | |
Western Blot 1:5 000 - 1:50000, Flow Cytometry, ELISA, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 1:2 000 - 1:20 000, Immunohistochemistry-Paraffin 1:20000 - 1:50000, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
0.2 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.2 ml distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
Antiserum with No Preservative | |
H3C14 | |
This Histone H3 antibody was raised against synthetic peptide derived from N terminal region of Human Histone H3, containing Citrulline at position 2;8;17 | |
Unpurified | |
RUO | |
Primary | |
This Histone H3 antibody reacts specifically with human Histone H3 Citrulline at either position 2 and or 8 and or 17. Not yet tested in other species, but the should react with a wide range of species due to sequence homology. | |
Human, Mouse, Rat, Bacteria | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction