Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Dimethyl Lys27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$548.00
Specifications
Classification | Polyclonal |
---|---|
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Research Discipline | Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls |
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot, Immunofluorescence.Specifications
Polyclonal | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
IgG | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Unconjugated | |
RUO | |
Human, Zebrafish | |
H3C14 | |
Primary | |
15 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title