Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Dimethyl Lys4 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB211022H
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Dot Blot.Specifications
Histone H3 (Dimethyl Lys4) | |
Polyclonal | |
Western Blot, Dot Blot | |
Q71DI3 | |
H3C14 | |
This Histone H3 [Dimethyl Lys4] antibody was raised against synthetic dimethylated peptide surrounding Lysine 4 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
0.05 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Human, Mouse, C. elegans | |
IgG |
Western Blot, Dot Blot | |
HRP | |
PBS with No Preservative | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction